6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR927220-100MG
OR927220-250MG
APOSOR927220-100MGEA
63.8
EUR
APOSOR927220-100MG
APOSOR927220-250MG
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 98%
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde
Formel:
C₈H₅BrN₂O Molecular Weight: 225.042 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09749988 CAS-Nummer: 1000341-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...