Fmoc-N-Me-Nle-OH 97%

Lieferant: Apollo Scientific

OR927345-1G OR927345-25G OR927345-5G
APOSOR927345-1GEA 0 EUR
APOSOR927345-1G APOSOR927345-25G APOSOR927345-5G
Fmoc-N-Me-Nle-OH 97%
Fmoc-N-Me-Nle-OH
Formel: C₂₂H₂₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00235878
CAS-Nummer: 112883-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR