7-Nitroisoquinolin-1(2H)-one 97%

Lieferant: Apollo Scientific

OR927390-1G
APOSOR927390-1GEA 213 EUR
APOSOR927390-1G
7-Nitroisoquinolin-1(2H)-one 97%
7-Nitroisoquinolin-1(2H)-one
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15526743
CAS-Nummer: 20141-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR