N-Methyl-3-(thien-2-yl)benzylamine 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-1-(3-thien-2-ylphenyl)methylamine

OR9274-1G
APOSOR9274-1GEA 395 EUR
APOSOR9274-1G
N-Methyl-3-(thien-2-yl)benzylamine 97%
N-Methyl-3-(thien-2-yl)benzylamine
Formel: C₁₂H₁₃NS
Molecular Weight: 203,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08435869
CAS-Nummer: 859833-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR