4-Azaspiro[2.4]heptan-5-one 95%

Lieferant: Apollo Scientific
Achtung

OR927431-1G OR927431-250MG
APOSOR927431-1GEA 510 EUR
APOSOR927431-1G APOSOR927431-250MG
4-Azaspiro[2.4]heptan-5-one 95%
4-Azaspiro[2.4]heptan-5-one
Formel: C₆H₉NO
Molecular Weight: 111.14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15146411
CAS-Nummer: 308266-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR