2-Ethyl-4,6-dihydroxypyrimidine 95%

Lieferant: Apollo Scientific

OR927450-1G
APOSOR927450-1GEA 36.4 EUR
APOSOR927450-1G
2-Ethyl-4,6-dihydroxypyrimidine 95%
2-Ethyl-4,6-dihydroxypyrimidine
Formel: C₆H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09907955
CAS-Nummer: 3709-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR