5-Chloro-2-(methylsulphonyl)pyrimidine 96%

Lieferant: Apollo Scientific
Achtung

OR927453-1G
APOSOR927453-1GEA 306 EUR
APOSOR927453-1G
5-Chloro-2-(methylsulphonyl)pyrimidine 96%
5-Chloro-2-(methylsulphonyl)pyrimidine
Formel: C₅H₅ClN₂O₂S
Molecular Weight: 192.623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00187881
CAS-Nummer: 38275-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR