5-(Dimethylamino)-pentanoic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR927457-250MG
APOSOR927457-250MGEA 27.7 EUR
APOSOR927457-250MG
5-(Dimethylamino)-pentanoic acid hydrochloride 95%
5-(Dimethylamino)-pentanoic acid hydrochloride
Formel: C₇H₁₅NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00035089
CAS-Nummer: 25726-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR