3,4,5-Trimethoxyphenol 95%

Lieferant: Apollo Scientific

Synonyme: Antiarol

OR927502-25G
APOSOR927502-25GEA 144 EUR
APOSOR927502-25G
3,4,5-Trimethoxyphenol 95%
3,4,5-Trimethoxyphenol
Formel: C₉H₁₂O₄
Molecular Weight: 184.19 g/mol
Siedepunkt: 283
Schmelzpunkt: 147
Lagertemperatur: Raumtemperatur
MDL: MFCD00008389
CAS-Nummer: 642-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR