5-(2-Pyridyl)isoxazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR927554-1G
APOSOR927554-1GEA 487 EUR
APOSOR927554-1G
5-(2-Pyridyl)isoxazole-3-carboxylic acid 95%
5-(2-Pyridyl)isoxazole-3-carboxylic acid
Formel: C₉H₆N₂O₃
Molecular Weight: 190,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07377113
CAS-Nummer: 893638-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR