3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine

Lieferant: Apollo Scientific
Achtung

OR927621-100MG
APOSOR927621-100MGEA 71.2 EUR
APOSOR927621-100MG
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20487103
CAS-Nummer: 1346447-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR