2-(1-Piperidinyl)benzoic acid 97%

Lieferant: Apollo Scientific

OR927641-1G OR927641-5G
APOSOR927641-1GEA 255 EUR
APOSOR927641-1G APOSOR927641-5G
2-(1-Piperidinyl)benzoic acid 97%
2-(1-Piperidinyl)benzoic acid
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205.253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00275762
CAS-Nummer: 42093-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR