1-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanone 95%

Lieferant: Apollo Scientific

OR927673-1G
APOSOR927673-1GEA 478 EUR
APOSOR927673-1G
1-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanone 95%
1-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanone
Formel: C₇H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00461792
CAS-Nummer: 10199-63-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR