6-Chloro-1H-indole-2-carbaldehyde 95%
Lieferant: Apollo Scientific
OR927744-1G
APOSOR927744-1GEA
88.6
EUR
APOSOR927744-1G
6-Chloro-1H-indole-2-carbaldehyde 95%
6-Chloro-1H-indole-2-carbaldehyde
Formel:
C₉H₆ClNO Molecular Weight: 179,603 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040767 CAS-Nummer: 53590-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...