5-Ethylthiophene-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR927755-1G OR927755-250MG
APOSOR927755-1GEA 623 EUR
APOSOR927755-1G APOSOR927755-250MG
5-Ethylthiophene-3-carboxylic acid 95%
5-Ethylthiophene-3-carboxylic acid
Formel: C₇H₈O₂S
Molecular Weight: 156.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419849
CAS-Nummer: 19156-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR