3-(1,3-Dioxolan-2-yl)aniline 95%

Lieferant: Apollo Scientific

OR927763-1G
APOSOR927763-1GEA 52.2 EUR
APOSOR927763-1G
3-(1,3-Dioxolan-2-yl)aniline 95%
3-(1,3-Dioxolan-2-yl)aniline
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272236
CAS-Nummer: 6398-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR