4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-on 95%

Lieferant: Apollo Scientific

Synonyme: Vanillylideneacetone, Dehydrozingerone, 4-(4-Hydroxy-3-methoxyphenyl)but-3-en-2-one

OR927851-25G
APOSOR927851-25GEA 137 EUR
APOSOR927851-25G
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-on 95%
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-on
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00012210
CAS-Nummer: 1080-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR