N-Boc-L-adamantylglycine 97%

Lieferant: Apollo Scientific

Synonyme: (2S)-2-(1-Adamant-1-yl)-2-((2-butoxycarbonyl)amino)acetic acid, (2S)-2-(1-Adamantyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid

OR927865-250MG
APOSOR927865-250MGEA 369 EUR
APOSOR927865-250MG
N-Boc-L-adamantylglycine 97%
N-Boc-L-adamantylglycine
Formel: C₁₇H₂₇NO₄
Molecular Weight: 309,401 g/mol
Schmelzpunkt: 180…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09952623
CAS-Nummer: 361441-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR