N,N'-Dimethyl-1,4-butanediamine 95%
Lieferant: Apollo Scientific
Synonyme:
N1,N4-Dimethylbutane-1,4-diamine
OR927887-5G
APOSOR927887-5GEA
79.9
EUR
APOSOR927887-5G
N,N'-Dimethyl-1,4-butanediamine 95%
N,N'-Dimethyl-1,4-butanediamine
Formel:
C₆H₁₆N₂ Molecular Weight: 116.205 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039799 CAS-Nummer: 16011-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...