4-[1-(Dimethylamino)ethyl]benzaldehyde 95%

Lieferant: Apollo Scientific

OR927896-1G OR927896-5G
APOSOR927896-1GEA 219 EUR
APOSOR927896-1G APOSOR927896-5G
4-[1-(Dimethylamino)ethyl]benzaldehyde 95%
4-[1-(Dimethylamino)ethyl]benzaldehyde
Formel: C₁₁H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08059974
CAS-Nummer: 915922-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR