2-Naphthalenepropanoic acid 97%

Lieferant: Apollo Scientific

OR927985-5G
APOSOR927985-5GEA 174 EUR
APOSOR927985-5G
2-Naphthalenepropanoic acid 97%
2-Naphthalenepropanoic acid
Formel: C₁₃H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00092742
CAS-Nummer: 21658-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR