Di(2-pyridyl)keton 97+%
Lieferant: Apollo Scientific
OR928006-100G
OR928006-25G
APOSOR928006-100GEA
393
EUR
APOSOR928006-100G
APOSOR928006-25G
Di(2-pyridyl)keton 97+%
Di(2-pyridyl)keton
Formel:
C₁₁H₈N₂O Schmelzpunkt: 52…55 °C Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006288 CAS-Nummer: 19437-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...