2-(Morpholin-3-yl)acetic acid hydrochloride 97%

Lieferant: Apollo Scientific

OR928013-1G OR928013-250MG OR928013-500MG
APOSOR928013-1GEA 0 EUR
APOSOR928013-1G APOSOR928013-250MG APOSOR928013-500MG
2-(Morpholin-3-yl)acetic acid hydrochloride 97%
2-(Morpholin-3-yl)acetic acid hydrochloride
Formel: C₆H₁₂ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11506159
CAS-Nummer: 86967-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR