Methyl-5-oxoprolinat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl DL-pyroglutamate, 5-Oxo-pyrrolidine-2-carboxylic acid methyl ester, Methyl 5-oxopyrrolidine-2-carboxylate, Methyl 2-pyrrolidinone-5-carboxylate

OR928083-5G
APOSOR928083-5GEA 21.9 EUR
APOSOR928083-5G
Methyl-5-oxoprolinat 95%
Methyl-DL-pyroglutamate
Formel: C₆H₉NO₃
Molecular Weight: 143.14 g/mol
Siedepunkt: 133…135/1 mm °C
Schmelzpunkt: 21…23 °C
Dichte: 1.25
MDL: MFCD03844651
CAS-Nummer: 54571-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR