4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 95%
Lieferant: Apollo Scientific
OR928099-1G
APOSOR928099-1GEA
40.6
EUR
APOSOR928099-1G
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 95%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile
Formel:
C₈H₄ClN₃ Molecular Weight: 177,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11046237 CAS-Nummer: 920965-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...