Ethyl indoline-6-carboxylate 95%

Lieferant: Apollo Scientific

OR928132-1G OR928132-250MG
APOSOR928132-1GEA 142 EUR
APOSOR928132-1G APOSOR928132-250MG
Ethyl indoline-6-carboxylate 95%
Ethyl indoline-6-carboxylate
Formel: C₁₁H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09954776
CAS-Nummer: 350683-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR