4-Amino-5-(2-furyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-5-(furan-2-yl)-4H-1,2,4-triazole-3-thiol

OR928166-1G
APOSOR928166-1GEA 109 EUR
APOSOR928166-1G
4-Amino-5-(2-furyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-furyl)-4H-1,2,4-triazole-3-thiol
Formel: C₆H₆N₄OS
Molecular Weight: 182,2 g/mol
Schmelzpunkt: 205…206 °C
MDL: MFCD03044388
CAS-Nummer: 80809-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR