4-Bromo-6-methylpyridin-2-ol 95%

Lieferant: Apollo Scientific

OR928203-1G
APOSOR928203-1GEA 85.7 EUR
APOSOR928203-1G
4-Bromo-6-methylpyridin-2-ol 95%
4-Bromo-6-methylpyridin-2-ol
Formel: C₆H₆BrNO
Molecular Weight: 188,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16610030
CAS-Nummer: 865156-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR