(1-Cyclopenten-1-yloxy)(trimethyl)silan 95%

Lieferant: Apollo Scientific

Synonyme: (Cyclopent-1-en-1-yloxy)trimethylsilane, (1-Cyclopentenyloxy)trimethylsilane

OR928213-5G
APOSOR928213-5GEA 98.6 EUR
APOSOR928213-5G
(1-Cyclopenten-1-yloxy)(trimethyl)silan 95%
1-(Trimethylsilyloxy)cyclopentene
Formel: C₈H₁₆OSi
Molecular Weight: 156.30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001395
CAS-Nummer: 19980-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR