3-Cyano-6-hydroxy-2-methoxypyridine 95%

Lieferant: Apollo Scientific

OR928260-1G OR928260-5G
APOSOR928260-1GEA 219 EUR
APOSOR928260-1G APOSOR928260-5G
3-Cyano-6-hydroxy-2-methoxypyridine 95%
3-Cyano-6-hydroxy-2-methoxypyridine
Formel: C₇H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11044304
CAS-Nummer: 130747-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR