4-(Methoxymethyl)phenol 95%

Lieferant: Apollo Scientific

OR928265-1G OR928265-250MG
APOSOR928265-1GEA 227 EUR
APOSOR928265-1G APOSOR928265-250MG
4-(Methoxymethyl)phenol 95%
4-(Methoxymethyl)phenol
Formel: C₈H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00020186
CAS-Nummer: 5355-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR