Sulphachloropyrazine sodium 98%

Lieferant: Apollo Scientific

OR928389-1G OR928389-5G
APOSOR928389-1GEA 0 EUR
APOSOR928389-1G APOSOR928389-5G
Sulphachloropyrazine sodium 98%
Sulphachloropyrazine sodium
Formel: C₁₀H₈ClN₄NaO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06798154
CAS-Nummer: 71720-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR