2-Ethyl-5-nitroaniline 98%

Lieferant: Apollo Scientific

Synonyme: 2-Ethyl-5-nitrobenzenamine

OR928420-100G
APOSOR928420-100GEA 130 EUR
APOSOR928420-100G
2-Ethyl-5-nitroaniline 98%
2-Ethyl-5-nitroaniline
Formel: C₈H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00034056
CAS-Nummer: 20191-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR