2-Methyl-3-hexanon 95%
Lieferant: Apollo Scientific
OR928462-1G
OR928462-5G
APOSOR928462-1GEA
0
EUR
APOSOR928462-1G
APOSOR928462-5G
2-Methyl-3-hexanon 95%
2-Methyl-3-hexanon
Formel:
C₇H₁₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009399 CAS-Nummer: 7379-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...