3-(2-Piperidyl)-1-propanol hydrochloride 95%

Lieferant: Apollo Scientific

OR928464-1G OR928464-250MG
APOSOR928464-1GEA 632 EUR
APOSOR928464-1G APOSOR928464-250MG
3-(2-Piperidyl)-1-propanol hydrochloride 95%
3-(2-Piperidyl)-1-propanol hydrochloride
Formel: C₈H₁₇NO·ClH
Molecular Weight: 179.688 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573686
CAS-Nummer: 90226-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR