Ethyl benzimidazole-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1H-benzo[d]imidazole-4-carboxylate

OR928479-1G
APOSOR928479-1GEA 72.5 EUR
APOSOR928479-1G
Ethyl benzimidazole-4-carboxylate 95%
Ethyl benzimidazole-4-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18251681
CAS-Nummer: 167487-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR