6-(Piperidin-1-yl)pyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-6-(piperidin-1-yl)pyridine, 1-(6-Carboxypyridin-2-yl)piperidine, 6-(Piperidin-1-yl)picolinic acid

OR9285-1G OR9285-250MG
APOSOR9285-1GEA 323 EUR
APOSOR9285-1G APOSOR9285-250MG
6-(Piperidin-1-yl)pyridine-2-carboxylic acid 97%
6-(Piperidin-1-yl)pyridine-2-carboxylic acid
Formel: C₁₁H₁₄N₂O₂
Schmelzpunkt: 97,5…101.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08435860
CAS-Nummer: 868755-50-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR