3-(Chloromethyl)pyridazine hydrochloride

Lieferant: Apollo Scientific

OR928537-1G
APOSOR928537-1GEA 371 EUR
APOSOR928537-1G
3-(Chloromethyl)pyridazine hydrochloride
3-(Chloromethyl)pyridazine hydrochloride
Formel: C₅H₅ClN₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD19705404
CAS-Nummer: 27349-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR