N1,N2-Dimethylcyclohexane-1,2-diamine 96%

Lieferant: Apollo Scientific
Achtung

OR928538-5G
APOSOR928538-5GEA 48 EUR
APOSOR928538-5G
N1,N2-Dimethylcyclohexane-1,2-diamine 96%
N1,N2-Dimethylcyclohexane-1,2-diamine
Formel: C₈H₁₈N₂
Molecular Weight: 142,2419 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05664382
CAS-Nummer: 61798-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR