ɑ-Methylene-γ-butyrolactone 95%

Lieferant: Apollo Scientific
Achtung

OR928542-5G
APOSOR928542-5GEA 0 EUR
APOSOR928542-5G
ɑ-Methylene-γ-butyrolactone 95%
alpha-Methylene-gamma-butyrolactone
Formel: C₅H₆O₂
Dichte: 1,12 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00005407
CAS-Nummer: 547-65-9
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR