Ethyl 5-benzoxazolecarboxylate 95%

Lieferant: Apollo Scientific

OR928548-1G
APOSOR928548-1GEA 133 EUR
APOSOR928548-1G
Ethyl 5-benzoxazolecarboxylate 95%
Ethyl 5-benzoxazolecarboxylate
Formel: C₁₀H₉NO₃
Molecular Weight: 191.183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682802
CAS-Nummer: 1404370-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR