Methyl 5-(2-furyl)isoxazole-3-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-(furan-2-yl)isoxazole-3-carboxylate

OR928555-5G
APOSOR928555-5GEA 266 EUR
APOSOR928555-5G
Methyl 5-(2-furyl)isoxazole-3-carboxylate 98%
Methyl 5-(2-furyl)isoxazole-3-carboxylate
Formel: C₉H₇NO₄
Molecular Weight: 193.156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03778977
CAS-Nummer: 33545-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR