4-Methyl-cinnamaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-3-(p-Tolyl)acrylaldehyde
OR928560-10G
APOSOR928560-10GEA
23.2
EUR
APOSOR928560-10G
4-Methyl-cinnamaldehyde 95%
4-Methyl-cinnamaldehyde
Formel:
C₁₀H₁₀O Molecular Weight: 146.186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460295 CAS-Nummer: 56578-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...