6-(Morpholin-4-yl)pyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(6-Carboxypyridin-2-yl)morpholine, 6-(Morpholin-4-yl)picolinic acid

OR9286-250MG
APOSOR9286-250MGEA 46.4 EUR
APOSOR9286-250MG
6-(Morpholin-4-yl)pyridine-2-carboxylic acid 97%
6-(Morpholin-4-yl)pyridine-2-carboxylic acid
Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,214 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03965867
CAS-Nummer: 554405-17-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR