2-(3-Phenoxyphenyl)propanoic acid

Lieferant: Apollo Scientific
Achtung

OR928603-1G
APOSOR928603-1GEA 95.7 EUR
APOSOR928603-1G
2-(3-Phenoxyphenyl)propanoic acid
2-(3-Phenoxyphenyl)propanoic acid
Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00072027
CAS-Nummer: 29679-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR