4-Methyl-1H-imidazole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR928643-1G
APOSOR928643-1GEA 487 EUR
APOSOR928643-1G
4-Methyl-1H-imidazole-2-carbaldehyde 95%
4-Methyl-1H-imidazole-2-carbaldehyde
Formel: C₅H₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD05864565
CAS-Nummer: 113825-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR