6-Acetyl-2H-1,4-benzoxazin-3(4H)-on 95%

Lieferant: Apollo Scientific

Synonyme: 6-Acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one

OR928736-25G
APOSOR928736-25GEA 173 EUR
APOSOR928736-25G
6-Acetyl-2H-1,4-benzoxazin-3(4H)-on 95%
6-Acetyl-2H-1,4-benzoxazin-3(4H)-on
Formel: C₁₀H₉NO₃
Molecular Weight: 191.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321313
CAS-Nummer: 26518-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR