3-Bromo-4-chloro-1H-indole 95%

Lieferant: Apollo Scientific
Achtung

OR928747-1G
APOSOR928747-1GEA 1130 EUR
APOSOR928747-1G
3-Bromo-4-chloro-1H-indole 95%
3-Bromo-4-chloro-1H-indole
Formel: C₈H₅BrClN
Molecular Weight: 230,489 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11894316
CAS-Nummer: 1181332-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR