2-Bromo-1,3-cyclohexanedione 95%

Lieferant: Apollo Scientific

OR928750-2G
APOSOR928750-2GEA 574 EUR
APOSOR928750-2G
2-Bromo-1,3-cyclohexanedione 95%
2-Bromo-1,3-cyclohexanedione
Formel: C₆H₇BrO₂
Molecular Weight: 191,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00971951
CAS-Nummer: 60060-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR