2-Methylindazole-7-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-2H-indazole-7-carboxylic acid

OR928767-1G
APOSOR928767-1GEA 37.7 EUR
APOSOR928767-1G
2-Methylindazole-7-carboxylic acid 98%
2-Methylindazole-7-carboxylic acid
Formel: C₉H₈N₂O₂
Molecular Weight: 176,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15071445
CAS-Nummer: 1234615-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR